Learn More
Biomatik Corporation Pig Neuron-specific enolase, NSE ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41100
Encompass_Preferred
Pig Neuron-Specific Enolase, Nse Elisa Kit for detecting Neuron-Specific Enolase, Nse in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9441