Learn More
Biomatik Corporation Human cluster of differentiation 34 (CD34) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41106
Encompass_Preferred
The Human cluster of differentiation 34 (CD34) ELISA Kit allows for quantitative determination of CD34 molecule in serum, plasma and tissue homogenates. Species Reactivity: Human. Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P28906. Restriction: For Research Use Only.
Catalog No. 50-149-9443