Learn More
Biomatik Corporation Human Tubulin Beta-3 Chain (TUBB3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41111
Encompass_Preferred
Human Tubulin Beta-3 Chain (Tubb3) Elisa Kit for detecting Tubulin, Beta 3 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13509; TUBB3. Target Synonyms: beta 3 tubulin; beta 4; beta-4; CDCBM; CDCBM1; CFEOM3; CFEOM3A; FEOM3; M (beta)3; M (beta)6; MC1R; Neuron specific beta III Tubulin; Neuron-specific class III beta-tubulin; QccE-11995; QccE-15186; TBB3_HUMAN; Tubb 3; TUBB3; TUBB4; Tubulin beta 3; Tubulin beta 3 chain; Tubulin beta 4; Tubulin beta III; Tubulin beta-3 chain; Tubulin beta-4 chain; Tubulin beta-III; tuj 1; tuj1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9448