Learn More
Biomatik Corporation Human Coagulation Factor XIII A1 Polypeptide (F13A1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41136
Encompass_Preferred
The Human Coagulation Factor XIII A1 Polypeptide (F13A1) ELISA Kit allows for quantitative determination of coagulation factor XIII, A1 polypeptide in serum, plasma. Species Reactivity: Human. Detection Range: 0.625 pg/mL-40 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P00488. Restriction: For Research Use Only.
Catalog No. 50-149-9456