Learn More
Biomatik Corporation Mouse Klotho ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41143
Encompass_Preferred
Mouse Klotho Elisa Kit for detecting Klotho in serum, plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O35082; Kl. Target Synonyms: Kl; Klotho; EC 3.2.1.31) [Cleaved into: Klotho peptide]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9458