Learn More
Biomatik Corporation Mouse hepcidin (Hepc) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41150
Encompass_Preferred
Mouse Hepcidin (Hepc) Elisa Kit for detecting Hepcidin Antimicrobial Peptide in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9EQ21; Hamp. Target Synonyms: Hamp; Hamp1; Hepc; Hepc1; Hepcidin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9460