Learn More
Biomatik Corporation Pig Trypsin ELISA Kit, 96 tests
Catalog No. 501499463
Encompass_Preferred
Pig Trypsin Elisa Kit for detecting Trypsin in serum, plasma. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9463 Supplier Biomatik Corporation Supplier No. EKC41173
Pig Trypsin Elisa Kit for detecting Trypsin in serum, plasma. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.