Learn More
Biomatik Corporation Chicken thyroxine (T4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41176
Encompass_Preferred
The Chicken thyroxine (T4) ELISA Kit allows for quantitative determination of thyroxine (T4) in serum, plasma. Species Reactivity: Chicken. Detection Range: 20 ng/mL-320 ng/mL. Sensitivity: 20 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9465