Learn More
Biomatik Corporation Pig prolactin (PRL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41178
Encompass_Preferred
Pig Prolactin (Prl) Elisa Kit for detecting Prolactin in serum, plasma. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01238; PRL. Target Synonyms: PRL; Prolactin; PRL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9466