Learn More
Biomatik Corporation Human Interleukin17C (IL17C) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41197
Encompass_Preferred
The Human Interleukin17C (IL17C) ELISA Kit allows for quantitative determination interleukin 17C in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 62.5 pg/mL-4000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9P0M4. Restriction: For Research Use Only.
Catalog No. 50-149-9470