Learn More
Biomatik Corporation Human 4-Hydroxynonenal (HNE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41207
Encompass_Preferred
The Human 4-Hydroxynonenal (HNE) ELISA Kit allows for quantitative determination of 4-Hydroxynonenal (HNE) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 39 pg/mL-2500 pg/mL. Sensitivity: 19.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9473