Learn More
Biomatik Corporation Human Aldose Reductase (AR) ELISA Kit, 96 tests
Catalog No. 501499474
Encompass_Preferred
The Human Aldose Reductase (AR) ELISA Kit allows for quantitative determination of aldo-keto reductase family 1, member B1 (aldose reductase) in serum, plasma, urine and tissue homogenates. Species Reactivity: Human. Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P15121. Restriction: For Research Use Only.
Catalog No. 50-149-9474 Supplier Biomatik Corporation Supplier No. EKC41209
The Human Aldose Reductase (AR) ELISA Kit allows for quantitative determination of aldo-keto reductase family 1, member B1 (aldose reductase) in serum, plasma, urine and tissue homogenates. Species Reactivity: Human. Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P15121. Restriction: For Research Use Only.