Learn More
Biomatik Corporation Pig Tumor necrosis factor (TNF/TNFA/TNFSF2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41247
Encompass_Preferred
Pig Tumor Necrosis Factor (Tnf/Tnfa/Tnfsf2) Elisa Kit for detecting Tumor Necrosis Factor (Tnf/Tnfa/Tnfsf2) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.047-3ng/ml. Sensitivity: 0.040ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P23563; TNF. Target Synonyms: TNF; TNFA; TNFSF2; Tumor necrosis factor; Cachectin; TNF-alpha; Tumor necrosis factor ligand superfamily member 2; TNF-a. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9484