Learn More
Biomatik Corporation Human bone morphogenetic protein 15 (BMP-15/GDF-9B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41301
Encompass_Preferred
The Human bone morphogenetic protein 15 (BMP-15/GDF-9B) ELISA Kit allows for quantitative determination of bone morphogenetic protein 15 in serum, plasma, cell culture supernates. Species Reactivity: Human. Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6PX77. Restriction: For Research Use Only.
Catalog No. 50-149-9497