Learn More
Biomatik Corporation Human Apolipoprotein M (APOM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41365
Encompass_Preferred
The Human Apolipoprotein M (APOM) ELISA Kit allows for quantitative determination of apolipoprotein M in serum, plasma, cell culture supernates. Species Reactivity: Human. Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O95445. Restriction: For Research Use Only.
Catalog No. 50-149-9514