Learn More
Biomatik Corporation Human C-C motif chemokine 26 (CCL26) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41416
Encompass_Preferred
The Human C-C motif chemokine 26 (CCL26) ELISA Kit allows for quantitative determination of chemokine (C-C motif) ligand 26 in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9Y258. Restriction: For Research Use Only.
Catalog No. 50-149-9521