Learn More
Biomatik Corporation Human CD109 antigen (CD109) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41419
Encompass_Preferred
The Human CD109 antigen (CD109) ELISA Kit allows for quantitative determination of CD109 molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 94 pg/mL-6000 pg/mL. Sensitivity: 23.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6YHK3. Restriction: For Research Use Only.
Catalog No. 50-149-9522