Learn More
Biomatik Corporation Human Properdin (CFP) ELISA Kit, 96 tests
Catalog No. 501499527
Encompass_Preferred
The Human Properdin (CFP) ELISA Kit allows for quantitative determination of complement factor properdin in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 12.5 ng/mL-800 ng/mL. Sensitivity: 3.12 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P27918. Restriction: For Research Use Only.
Catalog No. 50-149-9527 Supplier Biomatik Corporation Supplier No. EKC41443
The Human Properdin (CFP) ELISA Kit allows for quantitative determination of complement factor properdin in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 12.5 ng/mL-800 ng/mL. Sensitivity: 3.12 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P27918. Restriction: For Research Use Only.