Learn More
Biomatik Corporation Human Beta-Ala-His dipeptidase (CNDP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41459
Encompass_Preferred
The Human Beta-Ala-His dipeptidase (CNDP1) ELISA Kit allows for quantitative determination of carnosine dipeptidase 1 (metallopeptidase M20 family) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.04 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q96KN2. Restriction: For Research Use Only.
Catalog No. 50-149-9530