Learn More
Biomatik Corporation Human epiregulin (EREG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41544
Encompass_Preferred
The Human epiregulin (EREG) ELISA Kit allows for quantitative determination of epiregulin in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O14944. Restriction: For Research Use Only.
Catalog No. 50-149-9550