Learn More
Biomatik Corporation Human Ficolin-2 (FCN2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41562
Encompass_Preferred
The Human Ficolin-2 (FCN2) ELISA Kit allows for quantitative determination of ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.25 ng/mL-16 ng/mL. Sensitivity: 0.06 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q15485. Restriction: For Research Use Only.
Catalog No. 50-149-9553