Learn More
Biomatik Corporation Mouse Fibrinogen-like protein 1 (FGL1) ELISA Kit, 96 tests
Catalog No. 501499558
Encompass_Preferred
Mouse Fibrinogen-Like Protein 1 (Fgl1) Elisa Kit for detecting Fibrinogen-Like 1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q71KU9; Fgl1. Target Synonyms: Fgl1; Mfire1; Fibrinogen-like protein 1; Fibrinogen-related protein 1; Mfrep-1; Liver fibrinogen-related protein-1; Mfire-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9558 Supplier Biomatik Corporation Supplier No. EKC41577
Mouse Fibrinogen-Like Protein 1 (Fgl1) Elisa Kit for detecting Fibrinogen-Like 1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q71KU9; Fgl1. Target Synonyms: Fgl1; Mfire1; Fibrinogen-like protein 1; Fibrinogen-related protein 1; Mfrep-1; Liver fibrinogen-related protein-1; Mfire-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.