Learn More
Biomatik Corporation Human Interleukin-36 alpha (IL1F6) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41683
Encompass_Preferred
The Human Interleukin-36 alpha (IL1F6) ELISA Kit allows for quantitative determination interleukin 1 family, member 6 (epsilon) in serum, plasma, tissue homogenates, cell culture supernates. Species Reactivity: Human. Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9UHA7. Restriction: For Research Use Only.
Catalog No. 50-149-9578