Learn More
Biomatik Corporation Human Integrin alpha-IIb (ITGA2B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41709
Encompass_Preferred
The Human Integrin alpha-IIb (ITGA2B) ELISA Kit allows for quantitative determination integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P08514. Restriction: For Research Use Only.
Catalog No. 50-149-9581