Learn More
Biomatik Corporation Human neuregulin-4 (NRG4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41815
Encompass_Preferred
The Human neuregulin-4 (NRG4) ELISA Kit allows for quantitative determination of neuregulin-4 (NRG4) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 125 pg/mL-8000 pg/mL. Sensitivity: 31.25 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8WWG1. Restriction: For Research Use Only.
Catalog No. 50-149-9596