Learn More
Biomatik Corporation Mouse Pro-opiomelanocortin (POMC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41845
Encompass_Preferred
Mouse Pro-Opiomelanocortin (Pomc) Elisa Kit for detecting Pro-Opiomelanocortin (Pomc) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.73-3000pg/ml. Sensitivity: 0.18pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P01193. Target Synonyms: Pomc; Pomc1Pro-opiomelanocortin; POMC; Corticotropin-lipotropin) [Cleaved into: NPP; Melanotropin gamma; Gamma-MSH); Corticotropin; Adrenocorticotropic hormone; ACTH); Melanocyte-stimulating hormone alpha; Alpha-MSH; Melanotropin alpha); Corticotropin-like intermediary peptide; CLIP); Lipotropin beta; Beta-LPH); Lipotropin gamma; Gamma-LPH); Melanocyte-stimulating hormone beta; Beta-MSH; Melanotropin beta); Beta-endorphin; Met-enkephalin]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9600