Learn More
Biomatik Corporation Human Protein S100-A4 (S100A4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41892
Encompass_Preferred
The Human Protein S100-A4 (S100A4) ELISA Kit allows for quantitative determination of S100 calcium binding protein A4 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 0.9 ng/mL-60 ng/mL. Sensitivity: 0.225 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P26447. Restriction: For Research Use Only.
Catalog No. 50-149-9608