Learn More
Biomatik Corporation Human syndecan 4 (SDC4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41904
Encompass_Preferred
Human Syndecan 4 (Sdc4) Elisa Kit for detecting Syndecan 4 in serum, plasma, cell lysates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P31431. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9612