Learn More
Biomatik Corporation Human Semaphorin-7A (SEMA7A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41909
Encompass_Preferred
Human Semaphorin-7A (Sema7A) Elisa Kit for detecting Semaphorin 7A, Gpi Membrane Anchor (John Milton Hagen Blood Group) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O75326; SEMA7A. Target Synonyms: CD 108; CD108; CD108 antigen; CDw108; H Sema K1; H Sema L; H-SEMA-K1; H-Sema-L; JMH; JMH blood group antigen; John Milton Hagen blood group H Sema K1; John Milton Hargen human blood group Ag; John-Milton-Hargen human blood group Ag; MGC126692; MGC126696; SEM7A_HUMAN; Sema domain immunoglobulin domain (Ig) and GPI membrane anchor 7A; Sema K1; Sema L; SEMA7A; SEMAK1; SEMAL; Semaphorin 7A GPI membrane anchor; Semaphorin K1; Semaphorin L; Semaphorin-7A; Semaphorin-K1; Semaphorin-L; Semaphorin7a. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9614