Learn More
Biomatik Corporation Human Protein Wnt-2 (WNT2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC42021
Encompass_Preferred
The Human Protein Wnt-2 (WNT2) ELISA Kit allows for quantitative determination of wingless-type MMTV integration site family member 2 in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.45 ng/mL-30 ng/mL. Sensitivity: 0.11 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P09544. Restriction: For Research Use Only.
Catalog No. 50-149-9634