Learn More
Biomatik Corporation Fish dopamine (DA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC42066
Encompass_Preferred
The Fish dopamine (DA) ELISA Kit allows for quantitative determination of dopamine (DA) in serum, plasma and tissue homogenates. Species Reactivity: Fish. Detection Range: 62.5 pg/mL-1000 pg/mL. Sensitivity: 62.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9642