Learn More
Biomatik Corporation Human COL4 (Collagen Type Ⅳ) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60151
Encompass_Preferred
The Human COL4 (Collagen Type Ⅳ) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 0.78-50 ng/mL. Sensitivity: 0.47 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-149-9774