Learn More
Biomatik Corporation Human PⅢNP (N-Terminal Procollagen Ⅲ Propeptide) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60156
Encompass_Preferred
The Human PⅢNP (N-Terminal Procollagen Ⅲ Propeptide) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 23.44-1500 pg/mL. Sensitivity: 14.06 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-149-9778