Learn More
Biomatik Corporation Human TIMP-1 (Tissue Inhibitors of Metalloproteinase 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60157
Encompass_Preferred
Human TIMP-1 (Tissue Inhibitors of Metalloproteinase 1) ELISA Kit for detecting TIMP-1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.16-10ng/ml. Sensitivity: 0.1ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P01033. Target Synonyms: TIMP1; CLGI; EPA; EPO; HCI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9779