Learn More
Biomatik Corporation Human ADAM8 (A Disintegrin And Metalloprotease 8) ELISA Kit, 96 tests
Catalog No. 501499809
Encompass_Preferred
The Human ADAM8 (A Disintegrin And Metalloprotease 8) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 62.5-4000 pg/mL. Sensitivity: 37.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P78325. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-149-9809 Supplier Biomatik Corporation Supplier No. EKE60198
The Human ADAM8 (A Disintegrin And Metalloprotease 8) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 62.5-4000 pg/mL. Sensitivity: 37.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P78325. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.