Learn More
Biomatik Corporation Human FABP4 (Fatty Acid Binding Protein 4, Adipocyte) ELISA Kit, 96 tests
Catalog No. 501499821
Encompass_Preferred
The Human FABP4 (Fatty Acid Binding Protein 4, Adipocyte) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 0.39-25 ng/mL. Sensitivity: 0.23 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P15090. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-149-9821 Supplier Biomatik Corporation Supplier No. EKE60216
The Human FABP4 (Fatty Acid Binding Protein 4, Adipocyte) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 0.39-25 ng/mL. Sensitivity: 0.23 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P15090. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.