Learn More
Biomatik Corporation Human FASN (Fatty Acid Synthase) ELISA Kit, 96 tests
Catalog No. 501499851
Encompass_Preferred
The Human FASN (Fatty Acid Synthase) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 0.31-20 ng/mL. Sensitivity: 0.19 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P49327. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-149-9851 Supplier Biomatik Corporation Supplier No. EKE60269
Add to Cart
Add to Cart
The Human FASN (Fatty Acid Synthase) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 0.31-20 ng/mL. Sensitivity: 0.19 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P49327. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.