Learn More
Biomatik Corporation Human TTN (Titin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60303
Encompass_Preferred
Human TTN (Titin) ELISA Kit for detecting TTN in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.16-10ng/ml. Sensitivity: 0.1ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q8WZ42. Target Synonyms: CMD1G; CMH9; CMPD4; EOMFC; HMERF; LGMD2J; MYLK5; TMD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9878