Learn More
Biomatik Corporation Human AP36 (Apelin 36) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60316
Encompass_Preferred
The Human AP36 (Apelin 36) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 46.88-3000 pg/mL. Sensitivity: 28.13 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive. Detection Method: Colorimetric. Assay Time: 2.5 hours. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-149-9888