Learn More
Biomatik Corporation Human LDHA (Lactate Dehydrogenase A) ELISA Kit, 96 tests
Catalog No. 501499927
Encompass_Preferred
The Human LDHA (Lactate Dehydrogenase A) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 0.78-50 ng/mL. Sensitivity: 0.47 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P00338. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-149-9927 Supplier Biomatik Corporation Supplier No. EKE60382
Add to Cart
Add to Cart
The Human LDHA (Lactate Dehydrogenase A) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 0.78-50 ng/mL. Sensitivity: 0.47 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P00338. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.