Learn More
Biomatik Corporation Human TLR9 (Toll-Like Receptor 9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60609
Encompass_Preferred
Human TLR9 (Toll-Like Receptor 9) ELISA Kit for detecting TLR9 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.16-10ng/ml. Sensitivity: 0.1ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q9NR96. Target Synonyms: CD289. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0086