Learn More
Biomatik Corporation Human NSE (Neuron Specific Enolase) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60659
Encompass_Preferred
The Human NSE (Neuron Specific Enolase) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 2.34-150 ng/mL. Sensitivity: 1.4 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P09104. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-150-0109