Learn More
Biomatik Corporation Human TFF3 (Trefoil Factor 3, Intestinal) ELISA Kit, 96 tests
Catalog No. 501500130
Encompass_Preferred
Human TFF3 (Trefoil Factor 3, Intestinal) ELISA Kit for detecting TFF3 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.13-5000pg/ml. Sensitivity: 46.88pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q07654. Target Synonyms: ITF; P1B; TFI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0130 Supplier Biomatik Corporation Supplier No. EKE60685
Add to Cart
Add to Cart
Human TFF3 (Trefoil Factor 3, Intestinal) ELISA Kit for detecting TFF3 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.13-5000pg/ml. Sensitivity: 46.88pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q07654. Target Synonyms: ITF; P1B; TFI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.