Learn More
Biomatik Corporation Human ERα (Estrogen Receptor Alpha) ELISA Kit, 96 tests
Catalog No. 501500138
Encompass_Preferred
The Human ERα (Estrogen Receptor Alpha) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 62.5-4000 pg/mL. Sensitivity: 37.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P03372. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-150-0138 Supplier Biomatik Corporation Supplier No. EKE60697
The Human ERα (Estrogen Receptor Alpha) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 62.5-4000 pg/mL. Sensitivity: 37.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P03372. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.