Learn More
Biomatik Corporation Human SPC (Pulmonary Surfactant Associated Protein C) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60781
Encompass_Preferred
Human SPC (Pulmonary Surfactant Associated Protein C) ELISA Kit for detecting SPC in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.31-20ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P11686. Target Synonyms: SFTPC ; BRICD6; PSP-C; SFTP2; SMDP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0173