Learn More
Biomatik Corporation Human RNASE3/ECP (Ribonuclease A3/Eosinophil Cationic Protein) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60838
Encompass_Preferred
Human RNASE3/ECP (Ribonuclease A3/Eosinophil Cationic Protein) ELISA Kit for detecting RNASE3, ECP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 39.06-2500pg/ml. Sensitivity: 23.44pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P12724. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0219