Learn More
Biomatik Corporation Human MMP-8 (Matrix Metalloproteinase 8) ELISA Kit, 96 tests
Catalog No. 501500251
Encompass_Preferred
The Human MMP-8 (Matrix Metalloproteinase 8) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 0.16-10 ng/mL. Sensitivity: 0.1 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P22894. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-150-0251 Supplier Biomatik Corporation Supplier No. EKE60874
The Human MMP-8 (Matrix Metalloproteinase 8) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 0.16-10 ng/mL. Sensitivity: 0.1 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P22894. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.