Learn More
Biomatik Corporation Human TSP-1 (Thrombospondin-1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60959
Encompass_Preferred
Human TSP-1 (Thrombospondin-1) ELISA Kit for detecting TSP-1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.81-500ng/ml. Sensitivity: 4.69ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P07996. Target Synonyms: THBS1 ; THBS-1; TSP; TSP1; Thrombospondin-1p180. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0316