Learn More
Biomatik Corporation Human VEGFR-2/KDR (Vascular Endothelial Growth Factor Receptor 2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60966
Encompass_Preferred
Human VEGFR-2/KDR (Vascular Endothelial Growth Factor Receptor 2) ELISA Kit for detecting VEGFR-2, KDR in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.13-5000pg/ml. Sensitivity: 46.88pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P35968. Target Synonyms: VEGFR2; CD309; FLK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0321