Learn More
Biomatik Corporation Human TGFI (Transforming Growth Factor Beta Induced Protein) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60996
Encompass_Preferred
Human TGFβI (Transforming Growth Factor Beta Induced Protein) ELISA Kit for detecting TGFβI in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.63-40ng/ml. Sensitivity: 0.38ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q15582. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0348